siRNA Details
| Gene Symbol: Brat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276287.1 | NP_001263216.1 | 8 | 1225 | ||
| NM_181066.3 | NP_851411.1 | 8 | 1127 | |||
| GenBank mRNA | AK043212.1 | - | 8 | 1114 | ||
| AK049321.1 | - | 7 | 1024 | |||
| AK084987.1 | - | 8 | 1116 | |||
| AK085091.1 | - | 8 | 1116 | |||
| AK142387.1 | - | 5 | 2383 | |||
| AK154448.1 | - | 8 | 1098 | |||
| AK170269.1 | - | 8 | 1121 | |||
| BC068301.1 | - | 8 | 1191 | 30061848 | ||

Brat1
Genomic Map