siRNA Details
| Gene Symbol: Ap5z1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172725.2 | NP_766313.2 | 10 | 1384 | ||
| GenBank mRNA | AK040924.1 | - | 8 | 936 | ||
| AK082759.1 | - | 2 | 211 | |||
| AK129135.1 | - | 7 | 2582 | |||
| AK146431.1 | - | 10 | 1262 | |||
| AK152402.1 | - | 10 | 1262 | |||
| AK153787.1 | - | 9 | 2903 | |||
| BC057548.1 | - | 10 | 1239 | |||
| BC144755.1 | - | 8 | 915 | |||

Ap5z1
Genomic Map