siRNA Details
| Gene Symbol: Radil | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289588.1 | NP_001276517.1 | 4 | 1191 | ||
| NM_178702.4 | NP_848817.2 | 4 | 1191 | |||
| GenBank mRNA | AK038570.1 | - | 4 | 1124 | ||
| AK052964.1 | - | 4 | 1102 | |||
| AK079539.1 | - | 3 | 626 | |||
| AK083410.1 | - | 4 | 1209 | |||
| AK154169.1 | - | 4 | 1131 | |||
| AK162917.1 | - | 4 | 1171 | |||
| AK173277.1 | - | 4 | 1155 | |||
| BC056483.1 | - | 4 | 1143 | 5696312 | ||

Radil
Genomic Map