siRNA Details
| Gene Symbol: Zfp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289589.1 | NP_001276518.1 | 5 | 1030 | ||
| NM_001289590.1 | NP_001276519.1 | 5 | 1100 | |||
| NM_177681.4 | NP_808349.1 | 4 | 934 | |||
| GenBank mRNA | AK049650.1 | - | 4 | 890 | ||
| BC053080.1 | - | 5 | 939 | 5702966 | ||
| BC059836.1 | - | 5 | 1011 | 5697949 | ||

Zfp12
Genomic Map