siRNA Details
| Gene Symbol: Fam221a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172216.1 | NP_001165687.1 | 3 | 522 | ||
| NM_172727.3 | NP_766315.1 | 3 | 522 | |||
| GenBank mRNA | AK052323.1 | - | 3 | 436 | ||
| AK140239.1 | - | 3 | 428 | |||
| BC055818.1 | - | 3 | 450 | 4487036 | ||

Fam221a
Genomic Map