siRNA Details
| Gene Symbol: Ccdc129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081665.1 | NP_001075134.1 | 11 | 1577 | ||
| GenBank mRNA | AK085190.1 | - | 7 | 1221 | ||
| BC116335.2 | - | 11 | 1577 | 40097026 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc129| Gene Symbol: Ccdc129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081665.1 | NP_001075134.1 | 11 | 1577 | ||
| GenBank mRNA | AK085190.1 | - | 7 | 1221 | ||
| BC116335.2 | - | 11 | 1577 | 40097026 | ||