siRNA Details
| Gene Symbol: Thnsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033929.2 | NP_001029101.1 | 7, 8 | 1169 | ||
| NM_178413.5 | NP_848500.3 | 8, 9 | 1237 | |||
| GenBank mRNA | AI156509.1 | - | 1, 2 | 32 | ||
| AK165463.1 | - | 7, 8 | 1135 | |||
| BC025604.1 | - | 8, 9 | 1200 | 5250319 | ||
| BC051244.1 | - | 7, 8 | 1131 | 4975661 | ||
| BQ042728.1 | - | 4, 5 | 518 | |||

Thnsl2
Genomic Map