siRNA Details
| Gene Symbol: Thnsl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033929.2 | NP_001029101.1 | 3 | 351 | ||
| NM_178413.5 | NP_848500.3 | 4 | 419 | |||
| GenBank mRNA | AK165463.1 | - | 3 | 317 | ||
| BC025604.1 | - | 4 | 382 | 5250319 | ||
| BC051244.1 | - | 3 | 313 | 4975661 | ||
| CA491740.1 | - | 3 | 384 | |||
| CB600010.1 | - | 4 | 389 | |||
| CX223060.1 | - | 3 | 351 | |||

Thnsl2
Genomic Map