siRNA Details
| Gene Symbol: C87436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243741.1 | NP_001230670.1 | 10 | 2435 | ||
| NM_001243742.1 | NP_001230671.1 | 10 | 2001 | |||
| NM_146170.4 | NP_666282.3 | 10 | 1942 | |||
| GenBank mRNA | AK047127.1 | - | 10 | 1943 | ||
| AK077503.1 | - | 10 | 2322 | |||
| AK170524.1 | - | 11 | 1833 | |||
| AK171511.1 | - | 11 | 1833 | |||
| BC025632.1 | - | 10 | 1909 | 5321669 | ||
| BC026926.1 | - | 11 | 2066 | 3586097 | ||
| BC031913.1 | - | 10 | 1955 | 5376246 | ||
| BU613064.1 | - | 6 | 660 | |||

C87436
Genomic Map