siRNA Details
| Gene Symbol: Tmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081111.2 | NP_001074580.1 | 10 | 2481 | ||
| GenBank mRNA | BC150931.1 | - | 10 | 2492 | 9087842 | |
| BE627586.1 | - | 2 | 231 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmf1| Gene Symbol: Tmf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081111.2 | NP_001074580.1 | 10 | 2481 | ||
| GenBank mRNA | BC150931.1 | - | 10 | 2492 | 9087842 | |
| BE627586.1 | - | 2 | 231 | |||