siRNA Details
| Gene Symbol: Frmd4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145148.2 | NP_660130.2 | 7 | 966 | ||
| GenBank mRNA | AB042027.1 | - | 7 | 889 | ||
| AF327856.1 | - | 6 | 589 | |||
| AF327857.1 | - | 5 | 376 | |||
| AK051779.1 | - | 7 | 838 | |||
| AK129262.1 | - | 6 | 441 | |||
| AK132280.1 | - | 7 | 830 | |||
| AK143978.1 | - | 6 | 588 | |||
| BC052390.1 | - | 7 | 835 | 5718942 | ||

Frmd4b
Genomic Map