siRNA Details
| Gene Symbol: Slc6a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178703.4 | NP_848818.1 | 3 | 475 | ||
| GenBank mRNA | AK042956.1 | - | 3 | 337 | ||
| AK047663.1 | - | 3 | 321 | |||
| AK052971.1 | - | 3 | 300 | |||
| AK053798.1 | - | 314 | ||||
| AK053831.1 | - | 3 | 322 | |||
| AK053883.1 | - | 3 | 297 | |||
| BC059080.1 | - | 3 | 326 | 6827121 | ||
| CB519886.1 | - | 3 | 475 | |||
| M92378.1 | - | 2 | 237 | |||

Slc6a1
Genomic Map