siRNA Details
| Gene Symbol: C1rl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181344.5 | NP_851989.3 | 6 | 1461 | ||
| GenBank mRNA | AF456428.1 | - | 5 | 1345 | ||
| AK133495.1 | - | 6 | 1440 | |||
| AK134207.1 | - | 6 | 1462 | |||
| AK162497.1 | - | 6 | 1415 | |||
| BC070472.1 | - | 6 | 1424 | 30627947 | ||

C1rl
Genomic Map