siRNA Details
| Gene Symbol: Gprc5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181444.5 | NP_852109.2 | 2 | 695 | ||
| GenBank mRNA | BC034063.1 | - | 1 | 222 | ||
| BC036173.1 | - | 2 | 719 | 4989149 | ||
| BC036174.1 | - | 1 | 499 | |||
| BC039217.1 | - | 2 | 671 | 4205657 | ||
More Information
Set Membership: |
|

Gprc5a
Genomic Map