siRNA Details
| Gene Symbol: Caprin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301351.1 | NP_001288280.1 | 18 | 2501 | ||
| NM_181541.4 | NP_853519.2 | 19 | 3161 | |||
| GenBank mRNA | AK129460.1 | - | 18 | 2414 | ||
| AK140744.1 | - | 19 | 3164 | |||
| BC025636.1 | - | 19 | 3117 | |||
| BC027523.1 | - | 1 | 627 | |||
| BC092093.1 | - | 5 | 783 | |||
| BC125373.1 | - | 19 | 3050 | 40129888 | ||
| BC125375.1 | - | 19 | 3050 | 40129890 | ||

Caprin2
Genomic Map