siRNA Details
| Gene Symbol: Gm4876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047082.1 | - | 1 | 320 | ||
| BC120768.1 | - | 1 | 296 | 40129691 | ||
| BC120770.1 | - | 1 | 299 | 40129693 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4876| Gene Symbol: Gm4876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK047082.1 | - | 1 | 320 | ||
| BC120768.1 | - | 1 | 296 | 40129691 | ||
| BC120770.1 | - | 1 | 299 | 40129693 | ||