siRNA Details
Gene Symbol: Zfp418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146179.2 | NP_666291.2 | 4 | 1910 | ||
GenBank mRNA | AK039147.1 | - | 4 | 1872 | ||
AK054428.1 | - | 4 | 1908 | |||
BC024761.1 | - | 4 | 1890 | 5363178 | ||
BC026490.1 | - | 4 | 1886 | 4504776 |
