siRNA Details
| Gene Symbol: Vmn2r55 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104645.1 | NP_001098115.1 | 2 | 383 | ||
| GenBank mRNA | AB919108.1 | - | 2 | 383 | ||
| Gene Symbol: Vmn2r54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081449.2 | NP_001074918.2 | 2 | 383 | ||
| Gene Symbol: Vmn2r56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104648.1 | NP_001098118.1 | 2 | 383 | ||
| GenBank mRNA | AB919109.1 | - | 2 | 383 | ||
More Information
Set Membership: |
|

Vmn2r55
Genomic Map