siRNA Details
Gene Symbol: BC049730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199150.1 | NP_954601.1 | 7 | 867 | ||
GenBank mRNA | BC049730.1 | - | 7 | 867 | 6770790 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC049730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199150.1 | NP_954601.1 | 7 | 867 | ||
GenBank mRNA | BC049730.1 | - | 7 | 867 | 6770790 |