siRNA Details
| Gene Symbol: Atp1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290469.1 | NP_001277398.1 | 2, 3 | 259 | ||
| GenBank mRNA | AK053751.1 | - | 2, 3 | 231 | ||
| BC020177.1 | - | 2, 3 | 217 | 4506376 | ||
| BC034645.1 | - | 2, 3 | 215 | 5357731 | ||
| BC037206.1 | - | 2, 3 | 214 | 5362423 | ||
| BC042894.1 | - | 2, 3 | 259 | 5364812 | ||

Atp1a3
Genomic Map