siRNA Details
| Gene Symbol: Atp1a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290469.1 | NP_001277398.1 | 9, 10 | 1370 | ||
| GenBank mRNA | AK053751.1 | - | 9, 10 | 1342 | ||
| BC020177.1 | - | 9, 10 | 1328 | 4506376 | ||
| BC027000.1 | - | 2, 3 | 390 | |||
| BC034645.1 | - | 9, 10 | 1326 | 5357731 | ||
| BC037206.1 | - | 9, 10 | 1325 | 5362423 | ||
| BC042894.1 | - | 9, 10 | 1370 | 5364812 | ||

Atp1a3
Genomic Map