siRNA Details
Gene Symbol: Zfp574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168506.1 | NP_001161978.1 | 2 | 1414 | ||
NM_175477.4 | NP_780686.1 | 2 | 1469 | |||
GenBank mRNA | AK042074.1 | - | 2 | 1432 | ||
AK053744.1 | - | 2 | 1355 | |||
AK143997.1 | - | 2 | 1415 | |||
AK170243.1 | - | 2 | 1320 | |||
BC021833.2 | - | 2 | 1414 | |||
BC054064.1 | - | 2 | 1370 | |||
BC059044.1 | - | 2 | 1340 | 6816316 |
