siRNA Details
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144922.2 | NP_659171.1 | 1, 2 | 381 | ||
NM_178089.2 | NP_835190.1 | 1, 2 | 362 | |||
GenBank mRNA | AK042090.1 | - | 1, 2 | 362 | ||
AK087545.1 | - | 1, 2 | 351 | |||
AK143666.1 | - | 1, 2 | 424 | |||
AK155604.1 | - | 1, 2 | 392 | |||
AK202662.1 | - | 13 | ||||
AK205010.1 | - | 13 | ||||
BC021506.1 | - | 1, 2 | 405 | 5347099 |
