siRNA Details
Gene Symbol: Hnrnpul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144922.2 | NP_659171.1 | 6 | 945 | ||
NM_178089.2 | NP_835190.1 | 6 | 926 | |||
GenBank mRNA | AK042090.1 | - | 6 | 926 | ||
AK087545.1 | - | 6 | 915 | |||
AK143666.1 | - | 6 | 988 | |||
AK155604.1 | - | 6 | 956 | |||
AK183136.1 | - | 1 | 18 | |||
AK192923.1 | - | 1 | 18 | |||
AK195535.1 | - | 2 | 177 | |||
BC021506.1 | - | 6 | 969 | 5347099 | ||
BC027844.1 | - | 3 | 238 | |||
BC078642.1 | - | 6 | 869 | 6329630 |
