siRNA Details
| Gene Symbol: Itpkc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181593.2 | NP_853624.1 | 1 | 890 | ||
| GenBank mRNA | AK033605.1 | - | 1 | 559 | ||
| AK051297.1 | - | 1 | 958 | |||
| AK154888.1 | - | 1 | 922 | |||
| BC013227.1 | - | 1 | 776 | |||
| BC053450.1 | - | 1 | 890 | 6508198 | ||

Itpkc
Genomic Map