siRNA Details
| Gene Symbol: Hipk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033315.2 | NP_001028487.2 | 2 | 997 | ||
| GenBank mRNA | AB354583.1 | - | 2 | 741 | ||
| AK133498.1 | - | 1 | 1514 | |||
| BC147572.1 | - | 1 | 157 | 9056562 | ||
| BC147577.1 | - | 1 | 157 | 9056567 | ||

Hipk4
Genomic Map