siRNA Details
| Gene Symbol: 4931406P16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172741.2 | NP_766329.1 | 8, 9 | 1886 | ||
| GenBank mRNA | AK076994.1 | - | 8, 9 | 1887 | ||
| AK129125.1 | - | 7, 8 | 864 | |||
| AK170198.1 | - | 9, 10 | 1102 | |||
| BC060233.1 | - | 8, 9 | 1687 | 6852905 | ||

4931406P16Rik
Genomic Map