siRNA Details
Gene Symbol: Zfp939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243021.1 | NP_001229950.1 | 5 | 574 | ||
GenBank mRNA | AK034698.1 | - | 5 | 572 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243021.1 | NP_001229950.1 | 5 | 574 | ||
GenBank mRNA | AK034698.1 | - | 5 | 572 |