siRNA Details
| Gene Symbol: Siglecf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271019.1 | NP_001257948.1 | 8 | 1471 | ||
| NM_145581.2 | NP_663556.1 | 9 | 1573 | |||
| GenBank mRNA | AF293371.1 | - | 9 | 1509 | ||
| AK087658.1 | - | 8 | 1636 | |||
| AK146203.1 | - | 8 | 1610 | |||
| AK155469.1 | - | 9 | 1522 | |||
| AY210401.1 | - | 8 | 1469 | |||
| BC145038.1 | - | 7 | 1402 | 9053586 | ||
| BC145987.1 | - | 8 | 1504 | 40131290 | ||

Siglecf
Genomic Map