siRNA Details
| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177694.6 | NP_808362.2 | 4, 5 | 350 | ||
| GenBank mRNA | AB125740.1 | - | 4, 5 | 350 | ||
| AB206762.1 | - | 4, 5 | 350 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ano5| Gene Symbol: Ano5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177694.6 | NP_808362.2 | 4, 5 | 350 | ||
| GenBank mRNA | AB125740.1 | - | 4, 5 | 350 | ||
| AB206762.1 | - | 4, 5 | 350 | |||