siRNA Details
| Gene Symbol: Mtmr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172742.2 | NP_766330.2 | 16 | 2297 | ||
| GenBank mRNA | AK028584.1 | - | 4 | 922 | ||
| AK037069.1 | - | 1 | 58 | |||
| AK082105.1 | - | 1 | 142 | |||
| BC025088.1 | - | 1 | 359 | |||
| BC041672.1 | - | 16 | 2269 | |||
| BC055074.1 | - | 14 | 1961 | 30052208 | ||
| BC058196.1 | - | 9 | 1399 | |||

Mtmr10
Genomic Map