siRNA Details
| Gene Symbol: Mtmr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172742.2 | NP_766330.2 | 16 | 2049 | ||
| GenBank mRNA | AK028584.1 | - | 4 | 674 | ||
| BC025088.1 | - | 1 | 111 | |||
| BC041672.1 | - | 16 | 2021 | |||
| BC055074.1 | - | 14 | 1713 | 30052208 | ||
| BC058196.1 | - | 9 | 1151 | |||

Mtmr10
Genomic Map