siRNA Details
| Gene Symbol: Lrrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146191.3 | NP_666303.3 | 12, 13 | 1860 | ||
| GenBank mRNA | AK051264.1 | - | 12, 13 | 1848 | ||
| AK086795.1 | - | 12, 13 | 1849 | |||
| AK129441.1 | - | 9, 10 | 1273 | |||
| AK147469.1 | - | 12, 13 | 1846 | |||
| BC072664.1 | - | 12, 13 | 1809 | 30543489 | ||

Lrrk1
Genomic Map