siRNA Details
| Gene Symbol: Lrrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146191.3 | NP_666303.3 | 22 | 3414 | ||
| GenBank mRNA | AK051264.1 | - | 22 | 3402 | ||
| AK147469.1 | - | 22 | 3400 | |||
| BC072664.1 | - | 22 | 3363 | 30543489 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrk1| Gene Symbol: Lrrk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146191.3 | NP_666303.3 | 22 | 3414 | ||
| GenBank mRNA | AK051264.1 | - | 22 | 3402 | ||
| AK147469.1 | - | 22 | 3400 | |||
| BC072664.1 | - | 22 | 3363 | 30543489 | ||