siRNA Details
| Gene Symbol: Picalm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252520.1 | NP_001239449.1 | 12, 13 | 1605 | ||
| NM_001252521.1 | NP_001239450.1 | 12, 13 | 1605 | |||
| NM_146194.4 | NP_666306.2 | 12, 13 | 1605 | |||
| GenBank mRNA | AK144960.1 | - | 12, 13 | 1346 | ||
| AK162360.1 | - | 12, 13 | 1298 | |||
| AK220291.1 | - | 12, 13 | 1321 | |||
| AY206701.1 | - | 12, 13 | 1289 | |||
| BC011470.1 | - | 12, 13 | 1315 | 2651109 | ||
| BC023843.1 | - | 12, 13 | 1353 | 5346041 | ||
| BC027116.1 | - | 9, 10 | 916 | |||

Picalm
Genomic Map