siRNA Details
Gene Symbol: Thumpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145585.2 | NP_663560.1 | 4 | 751 | ||
GenBank mRNA | AK032078.1 | - | 4 | 727 | ||
AK152912.1 | - | 4 | 724 | |||
AK178112.1 | - | 2 | 177 | |||
BC004776.1 | - | 4 | 718 | 3257051 |
