siRNA Details
Gene Symbol: Thumpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145585.2 | NP_663560.1 | 2 | 405 | ||
GenBank mRNA | AK032078.1 | - | 2 | 381 | ||
AK084107.1 | - | 2 | 375 | |||
AK152912.1 | - | 2 | 378 | |||
AK183558.1 | - | 1 | 125 | |||
BC004776.1 | - | 2 | 372 | 3257051 | ||
BY031969.1 | - | 2 | 405 |
