siRNA Details
Gene Symbol: Dcun1d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163703.1 | NP_001157175.1 | 3 | 760 | ||
NM_173408.3 | NP_775584.1 | 3 | 779 | |||
GenBank mRNA | AK029043.1 | - | 3 | 725 | ||
AK155160.1 | - | 3 | 761 | |||
BC030335.1 | - | 3 | 702 | 5401034 |
