siRNA Details
| Gene Symbol: Dcun1d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163703.1 | NP_001157175.1 | 3 | 1020 | ||
| NM_173408.3 | NP_775584.1 | 3 | 1039 | |||
| GenBank mRNA | AK029043.1 | - | 3 | 985 | ||
| AK155160.1 | - | 3 | 1021 | |||
| BC030335.1 | - | 3 | 962 | 5401034 | ||

Dcun1d3
Genomic Map