siRNA Details
| Gene Symbol: Vwa3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177697.3 | NP_808365.2 | 16, 17 | 1915 | ||
| GenBank mRNA | AK086942.1 | - | 15, 16 | 1513 | ||
| AK132732.1 | - | 16, 17 | 1916 | |||
| AK136898.1 | - | 15, 16 | 1514 | |||

Vwa3a
Genomic Map