siRNA Details
| Gene Symbol: Tnrc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144925.3 | NP_659174.3 | 12, 13 | 4224 | ||
| GenBank mRNA | AK147327.1 | - | 12, 13 | 3929 | ||
| BC005741.1 | - | 1, 2 | 59 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnrc6a| Gene Symbol: Tnrc6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144925.3 | NP_659174.3 | 12, 13 | 4224 | ||
| GenBank mRNA | AK147327.1 | - | 12, 13 | 3929 | ||
| BC005741.1 | - | 1, 2 | 59 | |||