siRNA Details
| Gene Symbol: D430042O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081022.1 | NP_001074491.1 | 16 | 2921 | ||
| GenBank mRNA | AK052492.1 | - | 3 | 920 | ||
| AK052528.1 | - | 11 | 2428 | |||
| BC030902.1 | - | 1 | 165 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D430042O09Rik| Gene Symbol: D430042O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081022.1 | NP_001074491.1 | 16 | 2921 | ||
| GenBank mRNA | AK052492.1 | - | 3 | 920 | ||
| AK052528.1 | - | 11 | 2428 | |||
| BC030902.1 | - | 1 | 165 | |||