siRNA Details
| Gene Symbol: D430042O09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081022.1 | NP_001074491.1 | 15 | 2505 | ||
| GenBank mRNA | AK052528.1 | - | 10 | 2012 | ||
| BC157963.1 | - | 14 | 2437 | |||
| BC158019.1 | - | 15 | 2493 | 9007325 | ||

D430042O09Rik
Genomic Map