siRNA Details
| Gene Symbol: Fus | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139149.2 | NP_631888.1 | 9 | 926 | ||
| GenBank mRNA | AF224264.1 | - | 9 | 851 | ||
| AK038226.1 | - | 7 | 3995 | |||
| AK084867.1 | - | 2 | 1708 | |||
| AK132141.1 | - | 919 | ||||
| AK139960.1 | - | 9 | 925 | |||
| AK145582.1 | - | 9 | 916 | |||
| AK146198.1 | - | 9 | 919 | |||
| AK149118.1 | - | 9 | 920 | |||
| BC040827.1 | - | 9 | 881 | 4503820 | ||
| BC058247.1 | - | 9 | 901 | 6314291 | ||

Fus
Genomic Map