siRNA Details
| Gene Symbol: 4933402N03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173409.4 | NP_775585.1 | 3 | 880 | ||
| GenBank mRNA | AK029607.1 | - | 911 | |||
| AK161407.1 | - | 3 | 911 | |||
| BC115646.1 | - | 3 | 880 | 40103091 | ||
| BC115647.1 | - | 3 | 880 | 40103092 | ||

4933402N03Rik
Genomic Map