siRNA Details
| Gene Symbol: Tpcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146206.4 | NP_666318.2 | 22, 23 | 1972 | ||
| GenBank mRNA | AK052930.1 | - | 22, 23 | 1973 | ||
| AK089490.1 | - | 22, 23 | 2036 | |||
| AK170863.1 | - | 21, 22 | 1918 | |||
| BC025890.1 | - | 5, 6 | 856 | |||
| BC043472.1 | - | 18, 19 | 1550 | |||
| BC069857.1 | - | 5, 6 | 660 | |||
| BC141195.1 | - | 22, 23 | 2056 | 9055807 | ||

Tpcn2
Genomic Map