siRNA Details
| Gene Symbol: Tpcn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146206.4 | NP_666318.2 | 8, 9 | 800 | ||
| GenBank mRNA | AK040854.1 | - | 8, 9 | 828 | ||
| AK052930.1 | - | 8, 9 | 801 | |||
| AK089490.1 | - | 8, 9 | 795 | |||
| AK090059.1 | - | 8, 9 | 803 | |||
| AK170863.1 | - | 8, 9 | 796 | |||
| BC043472.1 | - | 4, 5 | 378 | |||
| BC141195.1 | - | 8, 9 | 884 | 9055807 | ||

Tpcn2
Genomic Map