siRNA Details
| Gene Symbol: Erich1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034862.2 | NP_001030034.2 | 2 | 129 | ||
| GenBank mRNA | AK147828.1 | - | 2 | 104 | ||
| BY168161.1 | - | 2 | 129 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Erich1| Gene Symbol: Erich1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034862.2 | NP_001030034.2 | 2 | 129 | ||
| GenBank mRNA | AK147828.1 | - | 2 | 104 | ||
| BY168161.1 | - | 2 | 129 | |||