siRNA Details
| Gene Symbol: Arhgef10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037736.2 | NP_001032825.1 | 13 | 1576 | ||
| NM_172751.3 | NP_766339.2 | 14 | 1693 | |||
| GenBank mRNA | AK032452.1 | - | 12 | 1293 | ||
| AK054038.1 | - | 14 | 1630 | |||
| AK147669.1 | - | 14 | 1694 | |||
| AK220332.1 | - | 13 | 1501 | |||
| BC059212.1 | - | 9 | 858 | 6400425 | ||
| BC066074.1 | - | 13 | 1496 | 6855196 | ||

Arhgef10
Genomic Map