siRNA Details
Gene Symbol: Whsc1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079952.1 | - | 2 | 221 | ||
AK156746.1 | - | 5 | 1460 | |||
AK170040.1 | - | 5 | 1523 | |||
AK172238.1 | - | 5 | 1486 | |||
BC036336.1 | - | 4 | 1052 | |||
BC064447.1 | - | 5 | 1498 |
